Skip to Content
Merck
All Photos(1)

Key Documents

Z529575

Sigma-Aldrich

Silver Shield® gloves

size L

Synonym(s):

Antimicrobial Gloves

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504
NACRES:
NB.35

material

PE and EVOH

size

L

sterility

non-sterile

feature

disposable
latex free
powder-free

packaging

pkg of 10 pair

manufacturer/tradename

North SSG-11

L × thickness

16 in. (40.6 cm) × 2.7 mil

color

silver

Looking for similar products? Visit Product Comparison Guide

General description

High-tech Norfoil laminate provides a protective barrier from hazardous chemicals. These gloves provide excellent permeation protection against 280 hazardous chemicals and mixtures including: acetone, benzene, epoxy, methanol, MEK, sodium hydroxide, sulfuric acid (93%), toluene, and trichloroethylene.

Gloves feature a five-layer fabric compacted to a thickness of 2.7 mil. Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35°C. Each layer acts as a barrier against various epoxies and industrial chemicals.

  • Lightweight; reduces hand fatigue without sacrificing dexterity
  • Economical - Can be used as a disposable glove, without the need for cleaning and re-cycling
  • Does not contain latex or chemical accelerators - Little chance of allergic reactions
  • Ambidextrous, hypoallergenic
  • Can be used as a secondary inner glove - Where the dangers of mechanical damage to gloves are high

Legal Information

Silver Shield is a registered trademark of Siebe North, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service