Z529559
Silver Shield® gloves
size S
Synonym(s):
Antimicrobial Gloves, Silver-Infused Gloves
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
PE and EVOH
size
S
sterility
non-sterile
feature
disposable
latex free
powder-free
packaging
pkg of 10 pair
manufacturer/tradename
North SSG-7
L × thickness
16 in. (40.6 cm) × 2.7 mil
color
silver
Looking for similar products? Visit Product Comparison Guide
General description
High-tech Norfoil laminate provides a protective barrier from hazardous chemicals. These gloves provide excellent permeation protection against 280 hazardous chemicals and mixtures including: acetone, benzene, epoxy, methanol, MEK, sodium hydroxide, sulfuric acid (93%), toluene, and trichloroethylene.
Gloves feature a five-layer fabric compacted to a thickness of 2.7 mil. Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35°C. Each layer acts as a barrier against various epoxies and industrial chemicals.
Gloves feature a five-layer fabric compacted to a thickness of 2.7 mil. Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35°C. Each layer acts as a barrier against various epoxies and industrial chemicals.
- Lightweight; reduces hand fatigue without sacrificing dexterity
- Economical - Can be used as a disposable glove, without the need for cleaning and re-cycling
- Does not contain latex or chemical accelerators - Little chance of allergic reactions
- Ambidextrous, hypoallergenic
- Can be used as a secondary inner glove - Where the dangers of mechanical damage to gloves are high
Legal Information
Silver Shield is a registered trademark of Siebe North, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service