Key Documents
Z116408
Sigma-Aldrich
Double-ended needle
gauge 12, L 18 in.
About This Item
UNSPSC Code:
41122002
NACRES:
NB.43
packaging
pack of 6 ea
manufacturer/tradename
Cadence Science
L
18 in.
gauge
12
General description
304 SS. One noncoring tip, one flat-cut end.
Application
Double-ended needle has been used in the transfer of solution betweentwo flasks. It has also been used in a sequence alkylation reaction.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Dehydroalanine-containing peptides: preparation from phenylselenocysteine and utility in convergent ligation strategies
Matthew R, et al.
Nature Protocols, 3001?3010-3001?3010 (2006)
Ramón Guzmán-Mejía et al.
Nature protocols, 2(11), 2759-2766 (2007-11-17)
The use of microwave (MW) irradiation for the condensation reaction between acetophenone and alpha-phenylethylamine to prepare (R,R)-bis[alpha-phenylethyl]amine results in significantly reduced reaction times relative to the use of conventional heating. In this protocol, a secondary amine, (R,R)-bis(alpha-phenylethyl)amine is treated with