Key Documents
Z224170
Sigma-Aldrich
Chem-FLEX tubing clamp
for tubing o.d., 23/64 ‑ 23/64 in.
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
Pricing and availability is not currently available.
description
For tubing CHEM-FLEX 106, 125
feature
for tubing o.d. 23/64 ‑ 23/64 in.
General description
Noncorroding, lightweight nylon clamps secure CHEM-FLEX tubing to transfer needles and fittings.
Legal Information
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)