Key Documents
WHA77002805
Whatman® UNIFILTER® 96 well filter plates
0.45 μm, clear polystyrene, polypropylene membrane, non-sterile, biomolecule, 25 ea
Synonym(s):
Z746479, micro plate, multiwell plate, well plates
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Pricing and availability is not currently available.
Product Name
Whatman® UNIFILTER® plates, 96 well, well volume 800 μL, 0.45 μm polypropylene membrane
material
polypropylene
polypropylene membrane
sterility
non-sterile
feature
0.45 μm polypropylene membrane
packaging
pkg of 25 ea
manufacturer/tradename
Whatman 7700-2805
Whatman Article No. 28420808 (US reference)
size
96 well
well volume
800 μL
pore size
0.45 μm
binding type
non-treated surface
Features and Benefits
  • No cross talk. Patented integral filter design prevents well-to-well cross contamination.
  • Economical to use. Wide range of well volume options ensures efficient use of materials.
  • Better control. Choice of filter media allows control of the flow rates and retention characteristics.
  • Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)