Key Documents
WHA77002808
Whatman® UNIFILTER® 96 well filter plates
0.45 μm, clear polystyrene, cellulose acetate membrane, non-sterile, biomolecule, 25 ea
Synonym(s):
Z746487, micro plate, multiwell plate, well plates
About This Item
UNSPSC Code:
41104900
NACRES:
NB.22
Pricing and availability is not currently available.
Product Name
Whatman® UNIFILTER® plates, size 96 wells, well working volume 800 μL, 0.45 μm cellulose acetate membrane
material
clear polystyrene
conical bottom clear wells
polystyrene filter
sterility
non-sterile
feature
0.45 μm cellulose acetate membrane
packaging
pkg of 25 ea
manufacturer/tradename
Whatman 7700-2808
Whatman Article No. 28420810 (US reference)
maximum volume
800 μL
size
96 wells
well working volume
800 μL
pore size
0.45 μm
binding type
non-treated surface
Features and Benefits
  • No cross talk. Patented integral filter design prevents well-to-well cross contamination.
  • Economical to use. Wide range of well volume options ensures efficient use of materials.
  • Better control. Choice of filter media allows control of the flow rates and retention characteristics.
  • Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Marit van Lenthe et al.
Malaria journal, 18(1), 430-430 (2019-12-20)
Sulfadoxine-pyrimethamine (SP) is a cornerstone of malaria chemoprophylaxis and is considered for programmes in the Democratic Republic of Congo (DRC). However, SP efficacy is threatened by drug resistance, that is conferred by mutations in the dhfr and dhps genes. The