Skip to Content
Merck
All Photos(1)

Key Documents

W357308

Sigma-Aldrich

Methyl 2-methyl-3-furyl disulfide

≥98%, FG

Synonym(s):

2-methyl-3-furylmethyl disulfide, Methyl oxycyclosulfide 719

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8OS2
CAS Number:
Molecular Weight:
160.26
FEMA Number:
3573
EC Number:
Council of Europe no.:
11924
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.079
NACRES:
NA.21
Organoleptic:
meaty; vegetable; sulfurous
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.5600 (lit.)

bp

210 °C (lit.)

density

1.163 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; vegetable; sulfurous

SMILES string

CSSc1ccoc1C

InChI

1S/C6H8OS2/c1-5-6(9-8-2)3-4-7-5/h3-4H,1-2H3

InChI key

SRUTWBWLFKSTIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Natural occurrence: Cooked beef and tea.
Methyl 2-methyl-3-furyl disulfide is a sulfur-containing heterocyclic flavor compound that is reported to occur in freshly roasted in-shell peanuts.[1]

Application


  • Elucidating salt-reduction mechanisms of aroma-active compounds from yeast extracts through sensomics approaches and electroencephalography.: This study provides insights into how aroma-active compounds like Methyl 2-methyl-3-furyl disulfide can reduce the perceived saltiness in food products, using advanced sensomics methods combined with EEG analysis to map sensory responses (Shan et al., 2024).

  • Interactions between kafirin and pickle-like odorants in soy sauce flavor Baijiu: Aroma profile change and binding mechanism.: Research on the interaction of protein kafirin with Methyl 2-methyl-3-furyl disulfide in Baijiu, highlighting the compound′s role in altering the aroma profile through complex binding mechanisms (Zhu et al., 2023).

  • Key Odorants of Raw and Cooked Green Kohlrabi (Brassica oleracea var. gongylodes L.).: Identifies Methyl 2-methyl-3-furyl disulfide among key odorants contributing to the flavor of green kohlrabi, enhancing understanding of flavor compounds in vegetables (Marcinkowska et al., 2021).

  • Meaty aroma notes from free amino acids and thiamine in nitrite-reduced, dry-fermented, yeast-inoculated sausages.: Examines the formation of meaty aroma notes, including the role of Methyl 2-methyl-3-furyl disulfide, in fermented sausages, providing insights into meat flavor chemistry and potential applications in meat substitute products (Flores et al., 2021).

  • Characterization of Potent Odorants Causing a Pickle-like Off-Odor in Moutai-Aroma Type Baijiu by Comparative Aroma Extract Dilution Analysis, Quantitative Measurements, Aroma Addition, and Omission Studies.: This detailed study analyzes the impact of Methyl 2-methyl-3-furyl disulfide in creating specific off-odors in Baijiu, aiding in understanding and controlling flavor profiles in distillation products (Wang et al., 2020).


Biochem/physiol Actions

Odor at 1.0%
Taste at 150 ppb-0.1 ppm

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

183.9 °F - closed cup

Flash Point(C)

84.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Key aroma compounds in roasted in-shell peanuts.
Kaneko S, et al.
Bioscience, Biotechnology, and Biochemistry, 77(7), 1467-1473 (2013)
Mosciano, G.
Perfumer & Flavorist, 26, 68-68 (2001)
Lulu Wang et al.
Journal of agricultural and food chemistry, 68(6), 1666-1677 (2020-01-21)
A potent unpleasant aroma presenting a pickle-like off-odor in Moutai-aroma type Baijiu was studied by comparative aroma extract dilution analysis (AEDA). Two Moutai-aroma type Baijiu samples, one having no off-odor (A) and the other one presenting pickle-like off-odor (B), were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service