Skip to Content
Merck
All Photos(1)

Key Documents

W336203

Sigma-Aldrich

Methyl furfuryl disulfide

≥95%, FG

Synonym(s):

Furfuryl methyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8OS2
CAS Number:
Molecular Weight:
160.26
FEMA Number:
3362
EC Number:
Council of Europe no.:
11513
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.064
NACRES:
NA.21
Organoleptic:
coffee; meaty; alliaceous; roasted; sulfurous
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥95%

refractive index

n20/D 1.568 (lit.)
n20/D 1.568

bp

60-61 °C/0.8 mmHg (lit.)
60-61 °C/0.8 mmHg

density

1.175 g/mL at 20 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

coffee; meaty; alliaceous; roasted; sulfurous

SMILES string

CSSCc1ccco1

InChI

1S/C6H8OS2/c1-8-9-5-6-3-2-4-7-6/h2-4H,5H2,1H3

InChI key

CLSLQQCDHOZMDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 0.2-1.0 ppm

Other Notes

Natural occurrence: Coffee, malt, pork liver, wheat bread, asparagus, potato, roasted turkey, grilled beef, malt, cocoa, cooked rice, mushrooms, clams.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27, 70-70 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service