Key Documents
SYND211150
Synthware vacuum jacketd Vigreux distillation head
joint: ST/NS 14/20, column H 150 mm, immersion depth 25 mm
Synonym(s):
Synthware D211150, short path head, Vigreux distilling head
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D211150
column H
150 mm
immersion depth
25 mm
Related Categories
General description
Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. Vacuum adapter has an inner joint and a sealed in drip tip. To be used with a 25 mm immersion thermometer
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)