Skip to Content
Merck
All Photos(1)

Key Documents

SYND214200

Synthware vacuum jacketd Vigreux distillation head

joint: ST/NS 24/40, thermometer joint: ST/NS 10/18, column H 200 mm, immersion depth 25 mm

Synonym(s):

Synthware D214200, short path head, Vigreux distilling head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.51
Pricing and availability is not currently available.

Joint

thermometer joint: ST/NS10/18
joint: ST/NS24/40

packaging

pkg of 1 ea

manufacturer/tradename

Synthware D214200

column H

200 mm

immersion depth

25 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. Vacuum adapter has an inner joint and a sealed in drip tip. To be used with a 25 mm immersion thermometer

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service