BMSP7700T5M
5 mL Pipette Tips
pack of 100 ea
Synonym(s):
Pipette Tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
Recommended Products
material
colorless
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Benchmark Scientific P7700-T5M
maximum volume
5 mL
volume range
0.5 - 5 mL
binding type
non-treated surface
Related Categories
General description
Bulk pack of loose 5ml Pipette Tips.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service