Merck
Pipette Tips
ZipTip Pipette tips
Pipette tips are disposable plastic tips used on pipettors for accurate and precise dispensing of liquids. They are available in a variety of sizes and shapes to fit different types of pipettors and are used in basic laboratory research and clinical diagnostics.
Read More About
Products
Sort by Relevance
Showing 1-20 of 893
Change View
Showing 1-20 of 893
Compare
Product Number
Product Name
Product Description
Pricing
Eppendorf® epT.I.P.S. Pipette Tips
These pipette tips are available in standard Eppendorf Quality, Biopur®, PCR clean and low retention, and Dualfilter T.I.P.S. ® options, making them an excellent choice for applications in cell culture, pharmacology, microbiology, protein purification, genomics, or PCR.  
Eppendorf® Combitips Advanced® Pipette Tips
These repeater pipette tips come in variants: Eppendorf Quality, Biopur®, PCR clean, and forensic DNA grade. The Biopur® tips are suitable for applications where the presence of living microorganisms could potentially jeopardize the integrity of the experiment. Moreover, they are devoid of pyrogens, ensuring the absolute absence of endotoxins. This feature is essential for endeavors involving cell culture, pharmacology, and medical research. On the other hand, the PCR clean and forensic DNA grade tips are specifically designed for DNA extraction, sample processing, and polymerase chain reaction (PCR) setup.
BRAND® Pipette Tips
BRAND® pipette tips are colorless, available in both sterile and non-sterile versions, and made of polypropylene. They are produced using raw materials that do not contain di(2-hydroxyethyl)methyldodecylammonium (DiHEMDA) and Oleamid. These pipette tips are available in a volume range of 0.1 μl to 10 ml and can be autoclaved at a temperature of 121 °C. The Sterile tips are sanitized with beta radiation. Additionally, these pipette tips can be bought in bulk as racks.
Corning® Pipette Tips
These pipet tips are manufactured in a controlled molding environment to ensure accuracy and precision across all tip volumes. These pipette tips are nonpyrogenic, human DNA-free, RNase-/DNase-free, and provided in a sterile state.
ART® Self-Sealing Barrier Pipette Tips
Precision-molded tips are guaranteed to prevent carryover contamination during the aspiration and delivery of samples. All Art tips are certified RNase-, DNase-, DNA-, and pyrogen-free. Art tips are ideal for use in genetic studies, forensics, general PCR work, and radioisotope sampling. These pipette tips are available in a volume range of 10 μl to 5 ml. They are also available as Softfit for improved ergonomics and low retention. Additionally, they come with Solvent Safe carbon filters for ultimate pipettor protection when pipetting organic solvents.
Axygen Maxymum Recovery® Microvolume Pipette Tips
 These pipette tips are made from non-sterile polypropylene and are non-pyrogenic and autoclavable. They are compatible with a wide array of single and multi-channel pipettors. These tips are devoid of RNase and DNase and feature inner rings that ensure secure fitting, preventing any liquid leakage.
Corning® Axygen® Automation Tips
These pipette tips come in clear polypropylene and are offered in both sterile and non-sterile variations. They are completely free from RNase, DNase, and human gDNA. These tips are ideal for the precise and accurate dispensing of liquids for a range of automated liquid handling systems.
Finnpipette® Pipette Tips
The Finnpipette® tips are available in both clear and colored variants and are categorized as non-sterile pipette tips. These tips are known for their accuracy and precision in pipetting tasks and feature a hydrophobic, consistently smooth design, which prevents unwanted liquid retention.