Skip to Content
Merck
All Photos(2)

Key Documents

ALD00132

Sigma-Aldrich

4-Phenoxybenzenesulfonyl fluoride

95%

Synonym(s):

4-Phenoxybenzenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9FO3S
CAS Number:
Molecular Weight:
252.26
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

45-49 °C

SMILES string

FS(C1=CC=C(OC2=CC=CC=C2)C=C1)(=O)=O

InChI

1S/C12H9FO3S/c13-17(14,15)12-8-6-11(7-9-12)16-10-4-2-1-3-5-10/h1-9H

InChI key

PDXOBGGAZNQARB-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service