ALD00094
3-Methoxybenzenesulfonyl fluoride
Synonym(s):
3-Methoxybenzenesulfonyl fluoride
About This Item
Recommended Products
form
liquid
Quality Level
reaction suitability
reaction type: click chemistry
refractive index
n/D 1.5038
density
1.3071 at 25 °C
SMILES string
FS(C1=CC(OC)=CC=C1)(=O)=O
InChI
1S/C7H7FO3S/c1-11-6-3-2-4-7(5-6)12(8,9)10/h2-5H,1H3
InChI key
RGVNAVGZAZZJBS-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service