Skip to Content
Merck
All Photos(3)

Key Documents

776874

Sigma-Aldrich

K-Oxyma

97%

Synonym(s):

2-Cyano-2-(hydroxyimino)acetic acid ethyl ester, potassium salt, Ethyl (hydroxyimino)cyanoacetate potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5KN2O3
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: Addition Reactions

mp

141-147

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CCOC(/C(C#N)=N/[O-])=O.[K+]

InChI

1S/C5H6N2O3.K/c1-2-10-5(8)4(3-6)7-9;/h9H,2H2,1H3;/q;+1/p-1/b7-4+;

InChI key

ZFBMUWGNRJITLS-KQGICBIGSA-M

Application

  • Non-explosive replacement for HOBt and HOAt with at least comparable performance to HOAt
  • Outperforms HOAt in sterically demanding peptide bond formations
  • Compatible with carbodiimide-mediated; solution and solid phase peptide coupling;
  • Less epimerization than HOBt in fragment condensation reactions
  • K-salt especially useful in peptide coupling on mild acid-labile resins such as 2-chlorotrityl; leading to minimal to no cleavage of the peptide from the resin due to its more alkaline character
  • K-salt has a higher melting point than OxymaPure

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subiros-Funosas; R. et al
Chemistry (Weinheim An Der Bergstrasse, Germany), 15, 9394-9403 (2009)
Mona M Agwa et al.
International journal of biological macromolecules, 162, 246-261 (2020-06-13)
Alzheimer's disease (AD) is neurological disorder characterized by dementia which causes severe problems with behavior, thinking and memory. Systemic administration of therapeutics to the central nervous system (CNS) is usually associated with very low efficiency due to presence of blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service