Skip to Content
Merck
All Photos(1)

Key Documents

670693

Sigma-Aldrich

(−)-2,3-Bis[(2R,5R)-2,5-dimethylphospholano]maleic anhydride

98%

Synonym(s):

3,4-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]furan-2,5-dione, catASium® M(R)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24O3P2
CAS Number:
Molecular Weight:
326.31
Beilstein:
9428339
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Quality Level

Assay

≥98% (31P-NMR)
98%

SMILES string

C[C@@H]1CC[C@@H](C)P1C2=C(P3[C@H](C)CC[C@H]3C)C(=O)OC2=O

InChI

1S/C16H24O3P2/c1-9-5-6-10(2)20(9)13-14(16(18)19-15(13)17)21-11(3)7-8-12(21)4/h9-12H,5-8H2,1-4H3/t9-,10-,11-,12-/m1/s1

InChI key

OQHMFJKKVZGWDC-DDHJBXDOSA-N

General description

Sold in collaboration with Solvias AG

Application

(−)-2,3-Bis[(2R,5R)-2,5-dimethylphospholano]maleic anhydride can be used to prepare its rhodium metal complexes, which are used as catalysts in the asymmetric hydrogenation reactions of alkenes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

catASium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Synthesis of Potential Precursors of the HIV Drug MC1220 and Its Analogues by Hydrogenation of (1-Arylvinyl) pyrimidines
Loksha YM and Pedersen EB
Journal of Heterocyclic Chemistry, 55(6), 1352-1357 (2018)
Activation, Deactivation and Reversibility Phenomena in Homogeneous Catalysis: A Showcase Based on the Chemistry of Rhodium/Phosphine Catalysts
Alberico E, et al.
Catalysts, 9(7), 582-582 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service