Skip to Content
Merck
All Photos(1)

Documents

536814

Sigma-Aldrich

PAMAM-OH dendrimer, generation 5 solution

generation 5, 5% in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=5);dendri PAMAM(NHCH2CH2OH)128
Molecular Weight:
28950.86
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amidoethanol surface groups

form

liquid

mol wt

generation 5

feature

core type ethylenediamine core (2-carbon core)

concentration

5% in methanol

no. Surface Groups

128

refractive index

n20/D 1.3374

density

0.786 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 128 surface hydroxyl groups.

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service