Skip to Content
Merck
All Photos(1)

Documents

477834

Sigma-Aldrich

PAMAM-OH dendrimer, generation 2 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=2);dendri PAMAM(NHCH2CH2OH)16
Molecular Weight:
3271.93
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amidoethanol surface groups

form

liquid

mol wt

generation 2

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

16

refractive index

n20/D 1.364

bp

65 °C

density

0.862 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 16 surface hydroxyl groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service