Skip to Content
Merck
All Photos(2)

Key Documents

327522

Sigma-Aldrich

N-(4-Chlorophenyl)-1,2-phenylenediamine

97%

Synonym(s):

2-Amino-4′-chlorodiphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHC6H4NH2
CAS Number:
Molecular Weight:
218.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

117-119 °C (lit.)

functional group

chloro

SMILES string

Nc1ccccc1Nc2ccc(Cl)cc2

InChI

1S/C12H11ClN2/c13-9-5-7-10(8-6-9)15-12-4-2-1-3-11(12)14/h1-8,15H,14H2

InChI key

WEUBIWJPIRTWDF-UHFFFAOYSA-N

Application

N-(4-Chlorophenyl)-1,2-phenylenediamine (2-Amino-4′-chlorodiphenylamine) was used in preparation of N-(p-chloro)phenylbenzimidazolin-2-one and monoaminophenazine dyes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on benzimidazoles. Part VII. Kinetics and mechanism of the reaction of thiophenoxydehalogenation of NN'-disubstituted 2-chlorobenzimidazolium perchlorates
Dembech P, et al.
Journal of the Chemical Society B: Physical Organic, 557-560 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service