306061
Glycer(ol-d3)
99 atom % D
Synonym(s):
1,2,3-Propanetriol-d3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(DOCH2)2CHOD
CAS Number:
Molecular Weight:
95.11
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
vapor density
3.1 (vs air)
Quality Level
isotopic purity
99 atom % D
Assay
99% (CP)
autoignition temp.
698 °F
composition
D2O and H2O, ≤1.5%
refractive index
n20/D 1.4676 (lit.)
bp
182 °C (lit.)
mp
20 °C (lit.)
density
1.302 g/mL at 25 °C
mass shift
M+3
SMILES string
[2H]OCC(O[2H])CO[2H]
InChI
1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2/i4D,5D,6D
InChI key
PEDCQBHIVMGVHV-WVALGTIDSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service