Skip to Content
Merck
All Photos(1)

Key Documents

264784

Sigma-Aldrich

4,5-Dihydro-3(2H)-thiophenone

98%

Synonym(s):

3-Thiophanone, Tetrahydrothiophen-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6OS
CAS Number:
Molecular Weight:
102.15
Beilstein:
105201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.528 (lit.)

bp

175 °C (lit.)

density

1.194 g/mL at 25 °C (lit.)

functional group

ketone
thioether

SMILES string

O=C1CCSC1

InChI

1S/C4H6OS/c5-4-1-2-6-3-4/h1-3H2

InChI key

DSXFPRKPFJRPIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,5-Dihydro-3(2H)-thiophenone is a volatile organic sulfur compound (VOSC) produced by basidiomycetous yeasts[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Buzzini et al.
FEMS yeast research, 5(4-5), 379-385 (2005-02-05)
Thirty-seven basidiomycetous yeasts belonging to 30 species of seven genera were grown on media containing l-cysteine or l-methionine as sole nitrogen sources with the objective of evaluating volatile organic sulfur compound (VOSC) production. The headspace of yeast cultures was analyzed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service