Skip to Content
Merck
All Photos(1)

Key Documents

709980

Sigma-Aldrich

Ethyl methyl sulfone

for energy applications, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8SO2
CAS Number:
Molecular Weight:
108.16
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

solid

bp

239 °C (lit.)

mp

32-37 °C

density

1.1 g/cm3

application(s)

battery manufacturing

SMILES string

CCS(C)(=O)=O

InChI

1S/C3H8O2S/c1-3-6(2,4)5/h3H2,1-2H3

InChI key

YBJCDTIWNDBNTM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Andreas Heckmann et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(1), 156-163 (2016-11-20)
Batteries based on cathode materials that operate at high cathode potentials, such as LiNi
Toshihiko Mandai et al.
Physical chemistry chemical physics : PCCP, 21(23), 12100-12111 (2019-04-26)
To achieve a sustainable-energy society in the future, next-generation highly efficient energy storage technologies, particularly those based on multivalent metal negative electrodes, are urgently required to be developed. Magnesium rechargeable batteries (MRBs) are promising options owing to the many advantageous

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service