Skip to Content
Merck
All Photos(2)

Key Documents

151556

Sigma-Aldrich

2-(3-Chloropropyl)-2-methyl-1,3-dioxolane

97%

Synonym(s):

5-Chloro-2-pentanone ethylene ketal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13ClO2
CAS Number:
Molecular Weight:
164.63
Beilstein:
104617
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Assay

97%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

73-76 °C/7 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

CC1(CCCCl)OCCO1

InChI

1S/C7H13ClO2/c1-7(3-2-4-8)9-5-6-10-7/h2-6H2,1H3

InChI key

OFERIRWCHSOJJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(3-Chloropropyl)-2-methyl-1,3-dioxolane was used in the preparation of 7-[(4,4-Ethylenedioxy)pentoxy]-2H-1-benzopyran-2-one.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María C Gutiérrez et al.
Organic & biomolecular chemistry, 1(20), 3500-3506 (2003-11-06)
The first fluorogenic assay allowing for detection of microbial enzymes able to perform Baeyer-Villiger oxidation is described. This is based on the use of 4-oxopentyl umbelliferyl ether 1 as a fluorogenic substrate. When Baeyer-Villigerases active against this test ketone are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service