Skip to Content
Merck
All Photos(1)

Key Documents

24024AST

Supelco

Astec® CYCLOBOND I 2000 HP-RSP Chiral (5 μm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

TM=["CYCLOBOND"] I 2000 HP-RSP Chiral HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CYCLOBOND I 2000 HP-RSP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-50 °C temperature
172 bar pressure (2500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 4.6 mm

matrix

silica particle platform
fully porous particle

matrix active group

cyclodextrin, beta- phase

particle size

5 μm

pore size

100 Å

operating pH

3.5-7

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

In the design of this new chemistry, it was an objective to create a very stable and reproducible phase with shorter retention times, while maintaining or improving selectivity over CYCLOBOND I 2000 RSP. With that goal and more achieved, CYCLOBOND I 2000 HP-RSP separates by extended H-bonding capability, and offers broad chiral selectivity for chiral screening. It is most beneficial for basic and neutral compounds.

  • Bonded phase: (R,S)-Hydroxypropyl modified β-cyclodextrin (high performance)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Application

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indirect photodetection of pregnanolone on a Cyclobond column by high-performance liquid chromatography
Agnus, Benoit, et al.
Journal of Chromatography A, 633 (1), 27-33 (1994)
Rapid high-performance liquid chromatographic separation of barley malt a-amylase on cyclobond columns
Henson, Cynthia A., Stone, Julie M.
Journal of Chromatography A, 469, 361-367 (1989)
Assessment of the complexation degree of camptothecin derivatives and cyclodextrins using spectroscopic and separative methodologies
Foulon, C., et al.
Tetrahedron Asymmetry, 20 (21), 2482-2489 (2009)
Reprint of: Enantiomeric separation of functionalized ethano-bridged Troger bases using macrocyclic cyclofructan and cyclodextrin chiral selectors in high-performance liquid chromatography and capillary electrophoresis with application of principal component analysis
Weatherly, Choyce A., et al.
Journal of Chromatography. B, Biomedical Applications, 968, 40-48 (2014)
Roland Török et al.
Journal of chromatography. A, 1098(1-2), 75-81 (2005-11-30)
Direct high-performance liquid chromatographic methods were developed for the enantioseparation of (R,S)-bicalutamide (1) and its analogs (+/-)-3-chloro-N-(4-cyano-3-(trifluoromethyl)phenyl)-2-hydroxy-2-methylpropanamide (2), (+/-)-N-(4-cyano-3-(trifluoro-methyl)phenyl)-2-methyloxirane-2-carboxamide (3), (+/-)-4-fluorophenylsulfonyl-2-hydroxy-2-methylpropionic acid (4) and (+/-)-3-hydroxy-N-(4-cyano-3-(trifluoromethyl)phenyl)-2-hydroxy-2-methylpropanamide (5). The methods involved the use of a cellulose-based Chiralcel OD-H, macrocyclic glycopeptide-based Chirobiotic T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service