SRP3263
G-CSF human
Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
CSF-3, GM-CSF?, MGI-1G, pluripoietin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
10.0-25.0 ng/mL
mol wt
18.7 kDa
packaging
pkg of 10 μg
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CSF3(1440)
General description
Granulocyte colony stimulating factor (G-CSF) or colony stimulating factor 3 (CSF-3) is produced in response to specific stimulation by a variety of cells including macrophages, fibroblasts, endothelial cells and bone marrow stroma. Human and murine G-CSF are cross-species reactive. The gene encoding G-CSF is localized on human chromosome 17. Recombinant human G-CSF is an 18.7kDa protein consisting of 174 amino acid residues.
Biochem/physiol Actions
Granulocyte colony stimulating factor (G-CSF) or colony stimulating factor 3 (CSF-3) is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It controls the production, differentiation and function of granulocytes. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. The protein has been shown to have a role in bone repair in animal models and clinically approved for neutropenia treatment.
Physical form
Lyophilized from 5 mM Sodium Citrate, pH 4.0.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
H S Lu et al.
Archives of biochemistry and biophysics, 268(1), 81-92 (1989-01-01)
Molecular characteristics and secondary structures of recombinant methionyl human granulocyte colony stimulating factor produced by genetically engineered Escherichia coli are described. Limited radiolabeling of the protein with tritiated iodoacetate and determination of the labeled residue revealed that this recombinant protein
Frequent loss of heterozygosity on chromosome 17 at 17q11.2-q12 in Barrett's adenocarcinoma.
Swift A
British Journal of Cancer, 71(5), 995-998 (1995)
Administration of granulocyte colony-stimulating factor with radiotherapy promotes tumor growth by stimulating vascularization in tumor-bearing mice.
Kim JS
Oncology Reports, 34(1), 147-154 (2015)
Effect of Granulocyte-Colony Stimulating Factor on Endothelial Cells and Osteoblasts.
Liu XL
BioMed Research International, 8485721-8485721 (2016)
Jessy Arbez et al.
Cytotherapy, 17(7), 948-955 (2015-03-31)
This study aimed to characterize the immune effectors contained in the grafts from donor mice mobilized by granulocyte colony-stimulating factor (G-CSF) and plerixafor and to evaluate their impact on the development of acute graft-versus-host-disease (aGVHD). Mobilization was done with G-CSF
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service