Skip to Content
Merck
All Photos(1)

Key Documents

SRP3093

Sigma-Aldrich

IL-4 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

B cell stimulating factor (BSF-1), BCDF, BCGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.2 ng/mL ED50

mol wt

14.9 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL4(3565)

General description

IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival and gene expression. Produced by mast cells, T cells and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells, characterized by their cytokine-secretion profile that includes secretion of IL-4, IL-5, IL-6, IL-10, and IL-13, which favor a humoral immune response. Another dominant function of IL-4 is the regulation of immunoglobulin class switching to the IgG1 and IgE isotypes. Excessive IL-4 production by Th2 cells has been associated with elevated IgE production and allergy. Recombinant human IL-4 is a 14.9 kDa globular protein containing 129 amino acid residues.

Application

Interleukin-4 (IL-4) has been used as a stimulant of M2 state of microglia that is associated with wound healing and debris clearance.

Biochem/physiol Actions

IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival and gene expression. Recombinant human IL-4 is a 14.9 kDa globular protein containing 129 amino acid residues.

Sequence

MHKCDITLQE IIKTLNSLTE QKTLCTELTV TDIFAASKNT TEKETFCRAA TVLRQFYSHH EKDTRCLGAT AQQFHRHKQL IRFLKRLDRN LWGLAGLNSC PVKEANQSTL ENFLERLKTI MREKYSKCSS

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijie Li et al.
Reproductive biology and endocrinology : RB&E, 19(1), 128-128 (2021-08-26)
Endometriosis is a serious reproductive and general health consequences. Recombinant human IL-37 (rhIL-37) is an inhibitor of inflammation. ELISA assay was performed to detect the concentration of cytokines. Flow cytometry was used to analyze cell proportion. Besides, qRT-PCR and western
Interleukin-4: a prototypic immunoregulatory lymphokine.
W E Paul
Blood, 77(9), 1859-1870 (1991-05-01)
Lu Liu et al.
Journal of experimental & clinical cancer research : CR, 41(1), 358-358 (2022-12-25)
IFN-γ is a key mediator of tumor immunity that can induce macrophage polarization to suppress tumor growth. Cyclin G2 functions as a tumor suppressor in various cancer cells; however, its role in macrophages remains unclear. This study aimed to investigate
Tanima Chatterjee et al.
Cells, 12(12) (2023-06-28)
Chronic widespread pain (CWP) is associated with a high rate of disability and decreased quality of life in people with HIV-1 (PWH). We previously showed that PWH with CWP have increased hemolysis and elevated plasma levels of cell-free heme, which
Masanori Kobayashi et al.
Cancer immunology, immunotherapy : CII, 63(8), 797-806 (2014-04-30)
Dendritic cell (DC)-based cancer vaccines may have a significant benefit to patients with advanced pancreatic cancer. However, variations among clinical studies make it difficult to compare clinical outcomes. Here, we identified factors that determined the clinical benefits by analyzing data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service