Skip to Content
Merck
All Photos(3)

Key Documents

H2518

Sigma-Aldrich

Heneicosanol

≥98.5% (GC)

Synonym(s):

1-Heneicosanol, Heneicosyl Alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)20OH
CAS Number:
Molecular Weight:
312.57
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.5% (GC)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCO

InChI

1S/C21H44O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22/h22H,2-21H2,1H3

InChI key

FIPPFBHCBUDBRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sol Ji Choi et al.
Food chemistry, 204, 94-101 (2016-03-19)
Policosanol (PC) is a mixture of health promoting bioactive long-chain aliphatic alcohols. Here, we report that green tea (Camellia sinensis) leaves are the exceptionally rich plant-sources of PC. Young and tender leaves and old and turf leaves of C. sinensis
Ji-Eun Ra et al.
Food chemistry, 317, 126388-126388 (2020-02-23)
Policosanols is a health promoting aliphatic alcohol known as lipid-lowing agent. To enable maximising the functional properties of wheat, this research investigates the policosanol profiles and adenosine 5'-monophosphate-activated protein kinase (AMPK) activation potential of Korean wheat seedlings according to cultivars
Muthusamy Muthusamy et al.
Plants (Basel, Switzerland), 9(11) (2020-11-12)
The spectral quality and intensity of light, photoperiodism, and other environmental factors have profound impacts on the metabolic composition of light-dependent higher plants. Hence, we investigate the effects of fluorescent light (96 μmol m-2s-1) and white (100 μmol m-2s-1), blue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service