Skip to Content
Merck
All Photos(1)

Key Documents

8.22051

Sigma-Aldrich

1-Eicosanol

for synthesis

Synonym(s):

1-Eicosanol, Arachyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19OH
CAS Number:
Molecular Weight:
298.55
MDL number:
UNSPSC Code:
12352104
EC Index Number:
211-119-4
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

solid

autoignition temp.

245 °C

kinematic viscosity

8 cSt(80 °C)

mp

65 °C

transition temp

flash point 195 °C

density

0.911 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCCCCCCCCCCCCCC

InChI

1S/C20H42O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3

InChI key

BTFJIXJJCSYFAL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 62 °C
Melting range (upper value): ≤ 65 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

381.2 °F - open cup

Flash Point(C)

194 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service