Skip to Content
Merck
All Photos(1)

Key Documents

EHU058861

Sigma-Aldrich

MISSION® esiRNA

targeting human RIPK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGACTGACCCCTGCACAGACAGACCCCTTCCCCTCTCTGCGAAAGGACCAAGCCCCAGAAGTCACTCCATCTCCTACGGCTCGCAATTTCCAGAGGCCCCCTGGCACCTTCCAGCCTGATGTCGTGCGTCAAGTTATGGCCCAGCGGTGCCCCCGCCCCCTTGGTGTCCATCGAGGAACTGGAGAACCAGGAGCTCGTCGGCAAAGGCGGGTTCGGCACAGTGTTCCGGGCGCAACATAGGAAGTGGGGCTACGATGTGGCGGTCAAGATCGTAAACTCGAAGGCGATATCCAGGGAGGTCAAGGCCATGGCAAGTCTGGATAACGAATTCGTGCTGCGCCTAGAAGGGGTTATCGAGAAGGTGAACTGGGACCAAGATCCCAAGCCGGCTCTGGTGACTAAATTCATGGAGAACGGCTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Martin-Sanchez et al.
Scientific reports, 7, 41510-41510 (2017-02-01)
Iron deficiency has been associated with kidney injury. Deferasirox is an oral iron chelator used to treat blood transfusion-related iron overload. Nephrotoxicity is the most serious and common adverse effect of deferasirox and may present as an acute or chronic
Alessandra Pescatore et al.
Cell death & disease, 7(8), e2346-e2346 (2016-08-26)
Incontinentia Pigmenti (IP) is a rare X-linked disease characterized by early male lethality and multiple abnormalities in heterozygous females. IP is caused by NF-κB essential modulator (NEMO) mutations. The current mechanistic model suggests that NEMO functions as a crucial component
Yu Matsuzawa-Ishimoto et al.
The Journal of experimental medicine, 214(12), 3687-3705 (2017-11-02)
A variant of the autophagy gene
R S Al-Lamki et al.
Cell death & disease, 7(6), e2287-e2287 (2016-07-01)
We previously reported that renal clear cell carcinoma cells (RCC) express both tumor necrosis factor receptor (TNFR)-1 and -2, but that, in organ culture, a TNF mutein that only engages TNFR1, but not TNFR2, causes extensive cell death. Some RCC
Xinghui Li et al.
Immunity, 50(3), 576-590 (2019-02-17)
Elevated glucose metabolism in immune cells represents a hallmark feature of many inflammatory diseases, such as sepsis. However, the role of individual glucose metabolic pathways during immune cell activation and inflammation remains incompletely understood. Here, we demonstrate a previously unrecognized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service