Merck
  • Autophagy protein ATG16L1 prevents necroptosis in the intestinal epithelium.
Autophagy protein ATG16L1 prevents necroptosis in the intestinal epithelium.
The Journal of experimental medicine (2017-11-02)
Yu Matsuzawa-Ishimoto, Yusuke Shono, Luis E Gomez, Vanessa M Hubbard-Lucey, Michael Cammer, Jessica Neil, M Zahidunnabi Dewan, Sophia R Lieberman, Amina Lazrak, Jill M Marinis, Allison Beal, Philip A Harris, John Bertin, Chen Liu, Yi Ding, Marcel R M van den Brink, Ken Cadwell
ABSTRACT
A variant of the autophagy gene
Sigma-Aldrich
MISSION® esiRNA, targeting human MLKL
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Target shRNA Control Plasmid DNA, Targets no known genes from any species
Sigma-Aldrich
MISSION® esiRNA, targeting human RIPK3
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder