Skip to Content
Merck
All Photos(1)

Key Documents

57510

Sigma-Aldrich

Inosine 5′-monophosphate disodium salt hydrate

≥99.0% (HPLC)

Synonym(s):

5′-IMP-Na2, 5′-Inosinic acid disodium salt hydrate, I-5′-P, IMP, Inosinic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N4O8PNa2 · xH2O
CAS Number:
Molecular Weight:
392.17 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥99.0% (HPLC)

form

powder or crystals

optical activity

[α]20/D −32±2°, c = 1% in H2O

impurities

~25% water

SMILES string

[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H13N4O8P.2Na.H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

KQCIGEIXDXQSGM-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Inosine-5′-monophosphate (5′-IMP) may be used as a substrate to study the activity, specificity and kinetics of inosine 5′-monophosphate dehydrogenase (IMPDH) EC 1.1.1.205, the rate limiting enzyme for the formation of GMP and the guanine nucleotide pool.

Biochem/physiol Actions

Inosine monophosphate, or inosinic acid, is the ribonucleotide of hypoxanthine and the first nucleotide formed during purine synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inosine 5′-monophosphate dehydrogenase activity as a biomarker in the field of transplantation.
Glander P, Hambach P, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry (2011)
Tomonori Nakanishi et al.
International immunopharmacology, 10(1), 91-97 (2009-10-21)
Inosine 5'-monophosphate dehydrogenase (IMPDH) inhibition is a critical target in solid organ transplantation, and the development of mycophenolate mofetil (MMF) represents a major advance in transplant medicine. In this study, the in vitro and in vivo pharmacological effects of BMS-566419
Sofie Haglund et al.
Therapeutic drug monitoring, 33(2), 200-208 (2011-02-12)
There is a large interindividual variability in thiopurine metabolism. High concentrations of methylthioinosine-5'-monophosphate (meTIMP) and low concentrations of 6-thioguanine nucleotides (6-TGNs) have been associated with a lower response rate and an increased risk of adverse events. In this study, the
Chanporn Chaosap et al.
Food science of animal resources, 40(3), 338-349 (2020-05-20)
The effects of organic Zn-L-selenomethionine (Zn-L-SeMet) at 0.3 ppm on carcass composition, meat characteristics, fatty acid composition, glutathione peroxidase activity, and ribonucleotide content were compared against the commercial inorganic sodium selenite (Na-Se) and the combination of the two, in commercial
L Hedstrom et al.
Current medicinal chemistry, 18(13), 1909-1918 (2011-04-27)
Inosine 5'-monophosphate dehydrogenase (IMPDH) catalyzes the first committed step of guanosine 5'-monophosphate (GMP) biosynthesis, and thus regulates the guanine nucleotide pool, which in turn governs proliferation. Human IMPDHs are validated targets for immunosuppressive, antiviral and anticancer drugs, but as yet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service