Skip to Content
Merck
All Photos(1)

Key Documents

PHR1514

Supelco

Disodium Guanylate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Guanosine 5′-monophosphate disodium salt hydrate, 5′-GMP-Na2, 5′-Guanylic acid disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5Na2O8P · xH2O
CAS Number:
Molecular Weight:
407.18 (anhydrous basis)
Beilstein:
3586801
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1221000

API family

disodium guanylate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H14N5O8P.2Na.H2O/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(23-9)1-22-24(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H2,19,20,21)(H3,11,13,14,18);;;1H2/q;2*+1;/p-2/t3-,5-,6-,9-;;;/m1.../s1

InChI key

MEKWJTZTMVQYSU-CYCLDIHTSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Disodium Guanylate is a flavor enhancer, obtained via chemical synthesis. It is used in combination with monosodium glutamate and disodium inosinate and helps in contributing to a fundamental taste sensation called savory in both animal and vegetable foods.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3321 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Consumer's Dictionary of Food Additives, 7th Edition: Descriptions in Plain English of More Than 12,000 Ingredients Both Harmful and Desirable Found in Foods (2009)
Fenaroli's Handbook of Flavor Ingredients (2004)
Dictionary of Food ingredients (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service