Skip to Content
Merck
All Photos(1)

Key Documents

32301

Sigma-Aldrich

Ammonium acetate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021908
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

<0.001 hPa

Assay

≥98%

form

solid

impurities

≤0.005% KMnO4 red. matter (as O)
≤2% water (Karl Fischer)
≤5 mg/kg heavy metals (as Pb)

ign. residue

≤0.01%

pH

6.7-7.3 (25 °C, 5% in water)

mp

110-112 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤10 ppm
Cd: ≤2 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤2 ppm
Fe: ≤5 ppm
K: ≤50 ppm
Mg: ≤2 ppm
Mn: ≤5 ppm
Na: ≤50 ppm
Ni: ≤5 ppm
Pb: ≤2 ppm
Zn: ≤2 ppm

SMILES string

N.CC(O)=O

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium acetate is a colorless hygroscopic solid. It can be prepared by reacting glacial acetic acid with ammonia or ammonium carbonate.[1] It has been reported as potential inhibitor of Pd/C mediated hydrogenolysis of benzyl ether.[2]

Application

Ammonium acetate has been used in the mobile phase for the HPLC determination of ezetimibe and its metabolites.[3] It may be used in preparation of phenanthrimidazoles and retinimidazoles.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anima Ghosal et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(3), 314-320 (2004-02-24)
Ezetimibe [1-(4-fluorophenyl)-3(R)-[3-(4-fluorophenyl)-3(S)-hydroxypropyl]-4(S)-(4-hydroxyphenyl)-2-azetidinone] (Zetia; Schering-Plough, Kenilworth, NJ) is the first in a new class of cholesterol-lowering agents known as cholesterol absorption inhibitors. The objective of this study was to identify the isoform(s) of human liver and intestinal UDP-glucuronosyltransferase (UGT) enzymes responsible
Selective inhibition of benzyl ether hydrogenolysis with Pd/C due to the presence of ammonia, pyridine or ammonium acetate.
Sajiki H.
Tetrahedron Letters, 36(20), 3465-3468 (1995)
Reactions of phenanthraquinone and retenequinone with aldehydes and ammonium acetate in acetic acid solution1.
Steck EA and Day AR.
Journal of the American Chemical Society, 65(3), 452-456 (1943)
M Athar Masood et al.
Basic & clinical pharmacology & toxicology, 127(6), 533-550 (2020-11-03)
Urine drug testing (UDT) is an important analytical/bio-analytical technique that has inevitably become an integral and vital part of a testing programme for diagnostic purposes. This manuscript presents a tailor-made LC-MS/MS quantitative assay method development and validation for a custom

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service