Skip to Content
Merck
All Photos(5)

Key Documents

30315

Sigma-Aldrich

Potassium iodide

puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
99.0-100.5% (calc. to the dried substance)
form:
solid
Pricing and availability is not currently available.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 745 °C)

description

REDUCING SUBSTANCES complying

grade

puriss. p.a.

Assay

≥99.0%
99.0-100.5% (calc. to the dried substance)

form

solid

impurities

≤0.0005% heavy metals (as Pb)
≤0.001% total nitrogen (N)
≤0.005% thiosulfate (S2O3)
≤0.02% free alkali (as KOH)

loss

≤0.5% loss on drying, 105 °C

pH

6.0-8.0 (20 °C, 5%)

mp

681 °C (lit.)

anion traces

bromide, chloride (as Cl-): ≤100 mg/kg
iodate (IO3-): ≤2 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

As: ≤0.1 mg/kg
Ba: ≤20 mg/kg
Ca: ≤10 mg/kg
Fe: ≤3 mg/kg
Mg: ≤10 mg/kg
Na: ≤300 mg/kg

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium iodide (KI) is a chemical compound that can be used as an iodide ion source for nucleophilic displacement reactions. During the electrochemical oxidation of aldehydes, it acts as a mediator. In the presence of an acid, it can cleave some ethers. KI can iodinate aromatic and organothallium compounds in the presence of an oxidizing agent. Additionally, it can form symmetrical thiosulfonic S-esters by reacting with sulfonyl chlorides.

Application

Potassium iodide is used as:
  • An iodine source in the synthesis of 1-iodoalkynes and iodophenols from alkynes and phenols using tert-butyl hydroperoxide (TBHP) as oxidant.
  • A catalyst in the synthesis of sulfonated oxindoles by arylsulfonylation of activated alkenes using sulfonylhydrazides as sulfonyl precursor, and water as solvent.
  • An oxidative degradation inhibitor to reduces oxidative degradation of monoethanolamine (MEA).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nils Erik Halvorsen et al.
Pharmaceuticals (Basel, Switzerland), 13(2) (2020-02-15)
A simple color spot test for determining the presence of residual tetrabutylammonium (TBA) in various 18F-radiopharmaceuticals is described. The test can be performed in less than five minutes. Iodoplatinate-saturated TLC plates are initially spotted with the 18F-radiopharmaceutical to be tested
Y Miyachi et al.
The British journal of dermatology, 107(2), 209-214 (1982-08-01)
The effects of potassium iodide, colchicine and dapsone on the in vitro generation of polymorphonuclear leukocyte (PMN)-derived oxygen intermediates were investigated. These three drugs have beneficial effects on those conditions in which PMNs play an important pathogenetic role. Three oxygen
H B Li et al.
Journal of chromatography. A, 918(2), 335-339 (2001-06-16)
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a
V Witko-Sarsat et al.
Journal of immunology (Baltimore, Md. : 1950), 161(5), 2524-2532 (1998-09-02)
We previously demonstrated the presence of advanced oxidation protein products (AOPP), a novel marker of oxidative stress in the plasma of uremic patients receiving maintenance dialysis. The present study in a cohort of 162 uremic patients showed that plasma concentrations
Alumina-supported potassium iodide as a heterogeneous catalyst for biodiesel production from soybean oil.
Xie W and Li H.
J. Mol. Catal. A: Chem., 255(1), 1-9 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service