Skip to Content
Merck
All Photos(2)

Key Documents

91827

Supelco

Biotin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

color

white

mp

231-233 °C (lit.)

application(s)

clinical testing
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biotin is a bicyclic compound, which is widely distributed in food stuffs. It is essentially required as an important nutrient for animals.
Biotin is a water soluble vitamin, which functions as a coenzyme in several carboxylase-mediated metabolic reactions such as gluconeogenesis, biosynthesis of fatty acids, and metabolism of amino acids.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Biotin may be used as a certified reference material for the quantification of the analyte in food samples, feed, tablets and premixes using high-performance liquid chromatography/avidin binding and high-performance liquid chromatography with tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of biotin in feed, food, tablets, and premixes using HPLC?MS/MS
Holler U, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 831(1-2), 8-16 (2006)
Nutrient Requirements of Dogs and Cats (2006)
Determination of the biotin content of select foods using accurate and sensitive HPLC/avidin binding
Staggs GC, et al.
J. Food Compos. Anal., 17(6), 767-776 (2004)
Yuxi Feng et al.
PloS one, 6(2), e16865-e16865 (2011-03-08)
Vasoregression is a hallmark of vascular eye diseases but the mechanisms involved are still largely unknown. We have recently characterized a rat ciliopathy model which develops primary photoreceptor degeneration and secondary vasoregression. To improve the understanding of secondary vasoregression in
Sung Jae Shin et al.
Clinical and vaccine immunology : CVI, 15(8), 1277-1281 (2008-06-14)
We previously reported that protein antigens of serodiagnostic potential were more abundant in culture filtrates than cellular extracts from liquid cultures of Mycobacterium avium subsp. paratuberculosis (D. Cho and M. T. Collins, Clin. Vaccine Immunol. 13:1155-1161, 2006). Based on this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service