Skip to Content
Merck
All Photos(1)

Key Documents

2031

Sigma-Aldrich

D-(+)-Biotin

Synonym(s):

D-(+)-Biotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.43

Assay

≥97% (by assay)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL

cation traces

heavy metals: ≤0.001%

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)

InChI key

YBJHBAHKTGYVGT-UHFFFAOYSA-N

General description

A sulfur-containing vitamin that plays a role in numerous carboxylation and deamination reactions.
Growth factor that plays a role in numerous naturally occurring carboxylation reactions.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 2 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohd Anisul et al.
eLife, 10 (2021-08-18)
The virus SARS-CoV-2 can exploit biological vulnerabilities (e.g. host proteins) in susceptible hosts that predispose to the development of severe COVID-19. To identify host proteins that may contribute to the risk of severe COVID-19, we undertook proteome-wide genetic colocalisation tests
Paniz Izadi et al.
iScience, 24(8), 102822-102822 (2021-08-03)
The formation of combined electrogenic/electrotrophic biofilms from marine sediments for the development of microbial energy storage systems was studied. Sediment samples from the German coasts of the Baltic and the North Sea were used as inocula for biofilm formation. Anodic
Chun-Min Shan et al.
Cell reports, 35(7), 109137-109137 (2021-05-20)
Oncogenic histone lysine-to-methionine mutations block the methylation of their corresponding lysine residues on wild-type histones. One attractive model is that these mutations sequester histone methyltransferases, but genome-wide studies show that mutant histones and histone methyltransferases often do not colocalize. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service