Skip to Content
Merck
All Photos(1)

Key Documents

69154

Supelco

cis-8,11,14-Eicosatrienoic acid

analytical standard

Synonym(s):

Homo-γ-linolenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)3(CH2)6CO2H
CAS Number:
Molecular Weight:
306.48
Beilstein:
1796533
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

form

viscous liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.478 (lit.)

format

neat

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(O)=O

InChI

1S/C20H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13H,2-5,8,11,14-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-

InChI key

HOBAELRKJCKHQD-QNEBEIHSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

cis-8,11,14-Eicosatrienoic acid is an unsaturated fatty acid, present in various plant species including Echium Rauwolfii, Sisymbrium Irio, Rhytididelphus squarrosus (Hedw.) Warnst etc, which are of pharmaceutical interest.

Application

cis-8,11,14-Eicosatrienoic acid may be used as an analytical reference standard for the determination of the analyte in plant species and biological matrices by chromatography-based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of unsaturated fatty acids with a migrating double bond in complex biological matrices by gas chromatography with flame ionization and mass spectrometry detection
Ivankin AN, et al.
Journal of Analytical Chemistry, 71(11), 1131-1137 (2016)
Fatty acid composition of the cold-water-inhabiting freshwater red alga Sirodotia Kylin
Bhosale RA, et al.
Journal of Applied Phycology, 21(1), 99-102 (2009)
Fatty acid composition of wild edible mushrooms species: A comparative study
Ribeiro B, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 93(1), 29-35 (2009)
Chemical composition of essential oils from aerial parts of Sisymbrium Irio from Jordan
Al-Qudah MA and Abu ZMH
Journal of Chemistry, 7(1), 6-10 (2010)
Fatty Acid Pattern and alkaloids of Echium rauwolfii
Dawidar AM, et al.
International Journal of Science and Engineering Applications, 4(4), 208-213 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service