Skip to Content
Merck
All Photos(2)

Key Documents

23401

Supelco

Arachidonic acid

analytical standard

Synonym(s):

cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Immunocytophyte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4(CH=CHCH2)4CH2CH2CO2H
CAS Number:
Molecular Weight:
304.47
Beilstein:
1713889
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

Quality Level

grade

analytical standard

Assay

≥97.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.4872 (lit.)

bp

169-171 °C/0.15 mmHg (lit.)

mp

−49 °C (lit.)

density

0.922 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O

InChI

1S/C20H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-,16-15-

InChI key

YZXBAPSDXZZRGB-DOFZRALJSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Arachidonic acid for inflammation research studies: Research on gigantol, a natural compound, emphasized its anti-inflammatory properties. This study suggests a parallel with arachidonic acid′s known effects on inflammation pathways, providing a comprehensive view of its potential in pharmaceutical and biotechnological applications (Chowdhury et al., 2024).

  • High-purity arachidonic acid for biochemical assays: A novel study on Yinhua Miyanling tablets for treating ulcerative colitis employed metabolomics and network pharmacology, highlighting arachidonic acid′s crucial role in understanding biochemical pathways in therapeutic contexts (Wang et al., 2024).

  • Arachidonic acid as a signaling molecule in cell culture: A hepatoprotective study on cholic acid against liver injury used arachidonic acid for its significant roles in cellular signaling pathways, elucidating its mechanisms in medical research (Zhang et al., 2024).

Biochem/physiol Actions

Arachidonic acid (AA) is an unsaturated ω6 fatty acid constituent of the phospholipids of cell membranes. Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation. AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism. AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis. AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs). Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.
Arachidonic acid stimulates adhesion of MDA-MB-435 human metastatic cancer cells to extracellular matrix molecules (collagen IV and vitronectin) .

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuizhu Wang et al.
International journal of molecular sciences, 21(7) (2020-04-12)
Gastric ulcer (GU), a prevalent digestive disease, has a high incidence and is seriously harmful to human health. Finding a natural drug with a gastroprotective effect is needed. Ocotillol, the derivate of ocotillol-type saponins in the Panax genus, possesses good
Yongjian Yue et al.
Journal of cellular biochemistry, 120(8), 12300-12310 (2019-02-28)
The disorders of hemostasis and coagulation were believed to be the main contributors to the pathogenesis of pulmonary thromboembolism (PTE), and platelets are the basic factors regulating hemostasis and coagulation and play important roles in the process of thrombosis. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service