Skip to Content
Merck
All Photos(1)

Key Documents

33539-M

Sigma-Aldrich

2-Propanol

≥99.8% (GC), ACS reagent, reag. Ph. Eur., reag. ISO, USP/NF

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.8% (GC)
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

impurities

≤0.00017% free alkali (as NH3)
≤0.00034% peroxides (as H2O2)
≤0.0005% KMnO4 red. matter (as O)
≤0.00074% free acid (as C2H5COOH)
≤0.001% non-volatile matter
≤0.002% carbonyl compounds (as acetone)
≤0.002% carbonyl compounds (as propionaldehyde)
≤0.01% ethanol (GC)
≤0.1% methanol (GC)
≤0.1% water (Karl Fischer)

refractive index

n20/D 1.377 (lit.)
n20/D 1.3770-1.3780

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

H2O: in accordance

density

0.785 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.01 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Propanol (Isopropanol) is a secondary alcohol. It has been tested as a substitute to fuel for use in various fuel cells. CuO powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Suspension of 2-propanol with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with uniformly thick layer of P25-TiO2.This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process.

Application

2-Propanol may be employed as fuel in the polymer electrolyte membrane direct 2-propanol fuel cell.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariadelfina Molinaro et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1152, 122215-122215 (2020-07-03)
Recent studies report strategies for analysing immunosuppressive drugs in brain, liver and renal tissue, mostly in animals: we developed and validated a two steps combined enzymatic digestion/mass spectrometry assay to quantify Tacrolimus (TAC) in heart biopsies. Our aims were to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service