Skip to Content
Merck
All Photos(1)

Key Documents

24105-M

Sigma-Aldrich

Ethanol (denatured)

96.0-97.2%, puriss., meets analytical specification of Ph. Eur., BP

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21

product name

Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

grade

puriss.

Assay

96.0-97.2%

form

liquid

autoignition temp.

683 °F

quality

meets analytical specification of Ph. Eur., BP

expl. lim.

19 %, 60 °F

impurities

Residual solvents, complies
acid. or alk. react. impurities, in accordance
volatile impurities, in accordance
≤0.001% non-volatile matter
≤0.02% methanol (GC)
4.8-7.4% water (Karl Fischer)

refractive index

n20/D 1.3600 (lit.)

pH

7 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

density

0.805-0.812 g/mL at 20 °C

absorption

in accordance

suitability

complies for appearance
complies for identity A
complies for identity B

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Chemical Analysis of the Antihyperglycemic, and Pancreatic α-Amylase, Lipase, and Intestinal α-Glucosidase Inhibitory Activities of Cannabis sativa L. Seed Extracts.: This research uses ethanol as a solvent in the extraction process of Cannabis sativa seeds to evaluate its antihyperglycemic properties and enzymatic inhibition, which has significant implications for the development of therapeutic agents (Haddou et al., 2023).

  • GC/MS and 2D NMR-based approach to evaluate the chemical profile of hydroalcoholic extract from Agaricus blazei Murill and its anti-inflammatory effect on human neutrophils.: This study utilizes a hydroalcoholic solution, primarily ethanol, to extract and analyze the chemical constituents of Agaricus blazei Murill, highlighting its potential anti-inflammatory properties (Campelo et al., 2024).

  • Quantum Diamonds at the Beach: Chemical Insights into Silica Growth on Nanoscale Diamond using Multimodal Characterization and Simulation.: This research provides insights into the chemical interactions facilitated by ethanol in the growth processes of silica on nanoscale diamonds, showcasing the role of ethanol in nanotechnology and materials science (Sandoval et al., 2023).

  • Chemical Analysis of an Isotopically Labeled Molecule Using Two-Dimensional NMR Spectroscopy at 34 μT.: Ethanol is employed as a solvent in the chemical analysis of isotopically labeled molecules using NMR spectroscopy, emphasizing the versatility of ethanol in enhancing spectral resolution and analysis accuracy (Lee et al., 2023).

Caution

Pertains only in Germany: Nicht zu Fertigungszwecken1

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of thin film dye sensitized solar cells with solar to electric power conversion efficiency over 10%.
Ito S, et al.
Thin Solid Films, 516(14), 4613-4619 (2008)
Christophe Tastet et al.
Proteomics, 3(2), 111-121 (2003-02-26)
Several zwitterionic detergents differing in their polar heads, linker parts and hydrophobia tail were synthesized and evaluated for their efficiency in protein solubilizers for two-dimensional electrophoresis. A model system consisting of human red blood cell ghosts was used for this
Spectrophotometric characteristics of chlorophylls a and b and their pheophytins in ethanol.
J F Wintermans et al.
Biochimica et biophysica acta, 109(2), 448-453 (1965-11-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service