Skip to Content
Merck
All Photos(1)

Key Documents

528100

Sigma-Aldrich

PI-103

A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR.

Synonym(s):

PI-103, 3-(4-(4-Morpholinyl)pyrido[3ʹ,2ʹ:4,5]furo[3,2-d]pyrimidin-2-yl)phenol, mTOR Inhibitor V, PI 3-K Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N4O3
CAS Number:
Molecular Weight:
348.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N5(CCOCC5)c1nc(nc3c1[o]c4ncccc43)c2cc(ccc2)O

InChI

1S/C19H16N4O3/c24-13-4-1-3-12(11-13)17-21-15-14-5-2-6-20-19(14)26-16(15)18(22-17)23-7-9-25-10-8-23/h1-6,11,24H,7-10H2

InChI key

TUVCWJQQGGETHL-UHFFFAOYSA-N

General description

A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR (IC50 = 2, 8, 88, 48, 150, 26, 20, and 83 nM for DNA-PK, p110α, p110β, p110δ, p110γ, PI3-KC2β, mTORC1, and mTORC2, respectively). It inhibits ATR and ATM only at much higher concentrations (IC50 = 850 and 920 nM, respectively) and exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10 µM. Shown to effectively block PI3-K/Akt signaling and cell proliferation in glioma cell lines both in vitro and in vivo. A 10 mM (2 mg/574 µl) solution of PI-103 (Cat. No. 528101) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA-PK, PI3-K, and mTOR
Product competes with ATP.
Reversible: no
Target IC50: 2, 8, 88, 48, 150, 26, 20, and 83 nM for DNA-PK, p110α, p110β, p110delta;, p110γ, PI3-KC2β, mTORC1, and mTORC2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming may be required for complete solubilization.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi-Wen Fan et al.
Cancer cell, 9(5), 341-349 (2006-05-16)
The PI3 kinase family of lipid kinases promotes cell growth and survival by generating the second messenger phosphatidylinositol-3,4,5-trisphosphate. To define targets critical for cancers driven by activation of PI3 kinase, we screened a panel of potent and structurally diverse drug-like
Florence I Raynaud et al.
Cancer research, 67(12), 5840-5850 (2007-06-19)
Extensive evidence implicates activation of the lipid phosphatidylinositide 3-kinase (PI3K) pathway in the genesis and progression of various human cancers. PI3K inhibitors thus have considerable potential as molecular cancer therapeutics. Here, we detail the pharmacologic properties of a prototype of
Zachary A Knight et al.
Cell, 125(4), 733-747 (2006-05-02)
Phosphoinositide 3-kinases (PI3-Ks) are an important emerging class of drug targets, but the unique roles of PI3-K isoforms remain poorly defined. We describe here an approach to pharmacologically interrogate the PI3-K family. A chemically diverse panel of PI3-K inhibitors was
PtdIns(3,4)P2, Lamellipodin, and VASP coordinate actin dynamics during phagocytosis in macrophages.
Monta??o-Rend??n, et al.
The Journal of cell biology, 221 (2023)
Franz F Dressler et al.
Nature communications, 15(1), 4513-4513 (2024-05-28)
Urothelial bladder cancer (UC) has a wide tumor biological spectrum with challenging prognostic stratification and relevant therapy-associated morbidity. Most molecular classifications relate only indirectly to the therapeutically relevant protein level. We improve the pre-analytics of clinical samples for proteome analyses

Articles

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service