Skip to Content
Merck
All Photos(3)

Key Documents

571474

Sigma-Aldrich

3,5-Di-tert-butylbenzyl bromide

97%

Synonym(s):

1-(Bromomethyl)-3,5-bis(1,1-dimethylethyl)benzene, 1-Bromomethyl-3,5-di-tert-butylbenzene, a-Bromo-3,5-di-tert-butyltoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3CH2Br
CAS Number:
Molecular Weight:
283.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

functional group

bromo

SMILES string

CC(C)(C)c1cc(CBr)cc(c1)C(C)(C)C

InChI

1S/C15H23Br/c1-14(2,3)12-7-11(10-16)8-13(9-12)15(4,5)6/h7-9H,10H2,1-6H3

InChI key

SNRYBGHMHAJTTM-UHFFFAOYSA-N

Application

Reactant involved in:
  • Chemoselective trifluoromethylation
  • Aralkylation of 2-N-acetylguanine
  • Coupling reactions with terminal alkynes
  • Synthesis of near-IR solid-state fluorescent naphthooxazine dyes

Reactant involved in synthesis of biologically active molecules including:
  • Steroid sulfatase inhibitors
  • Short cationic antimicrobial peptides

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service