Skip to Content
Merck
All Photos(2)

Key Documents

436887

Sigma-Aldrich

4-Vinylbenzyl chloride

90%

Synonym(s):

4-(Chloromethyl)styrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4CH2Cl
CAS Number:
Molecular Weight:
152.62
Beilstein:
2204384
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

form

liquid

contains

500 ppm tert-butylcatechol as inhibitor

refractive index

n20/D 1.572 (lit.)

viscosity

1,830 cP(25 °C)(lit.)

bp

229 °C (lit.)

density

1.083 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCc1ccc(C=C)cc1

InChI

1S/C9H9Cl/c1-2-8-3-5-9(7-10)6-4-8/h2-6H,1,7H2

InChI key

ZRZHXNCATOYMJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Vinylbenzyl chloride (VBC) is commonly used as a monomer that belongs to the class of aryl chlorides and contains both a vinyl group (CH2=CH-) and a benzyl chloride group (C6H4CH2Cl) in its structure. It is used as a reactive building block in various polymerization reactions to synthesize different types of polymers with excellent thermal stability, chemical resistance and good film-forming properties. It is commonly utilized in the production of resins, coatings, adhesives, and ion-exchange resins. It offers versatility in polymer synthesis, allowing for the introduction of desired functionalities into polymers.

It is also:
  • A component of ion exchange resins, photo-resist polymers, cross-linkable fibers, coupling agents and electroconducting polymers.
  • Starter for various copolymer preparations.
  • Dual functional monomer. Readily derivatized by chloride displacement.

Application

4-Vinylbenzyl chloride can be used as a monomer to synthesize:
  • The hypercross-linked polymer for supercapacitor applications. Incorporation of 4-vinyl benzyl chloride contributes to the formation of a porous structure and specific surface area which facilitate efficient electrolyte ion diffusion and provide ample sites for charge storage, leading to enhanced electrochemical performance.
  • The polymeric thin films by the matrix-assisted pulsed laser evaporation (MAPLE) technique. The resulting polymeric thin films can exhibit non-linear optical effects such as optical second harmonic generation, electro-optic modulation, or optical frequency conversion. These properties make them suitable for various applications in non-linear optic devices.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of Materials, 16, 2648-2655 (2004)
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 43, 140-141 (2002)
Chemistry of Materials, 16, 2282-2286 (2004)
Liwei Qian et al.
Analytica chimica acta, 884, 97-105 (2015-06-16)
Structural stability of the template is one of the most important considerations during the preparation of protein imprinting technology. To address this limitation, we propose a novel and versatile strategy of utilizing macromolecularly functional monomers to imprint biomacromolecules. Results from
Juanjuan Feng et al.
Analytical and bioanalytical chemistry, 407(23), 7025-7035 (2015-07-30)
Ionic liquids (ILs) have been efficiently used as a "designer sorbent" in sample preparation. A novel 1-(3-aminopropyl)-3-(4-vinylbenzyl)imidazolium 4-styrenesulfonate IL monomer was synthesized and copolymerized with 1,6-di(3-vinylimidazolium) hexane bishexafluorophosphate IL as cross-linking agent to prepare a cross-linked polymeric ionic liquids (PILs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service