Skip to Content
Merck
All Photos(1)

Documents

332402

Sigma-Aldrich

2-Mesitylmagnesium bromide solution

1.0 M in diethyl ether

Synonym(s):

2,4,6-Trimethylphenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2MgBr
CAS Number:
Molecular Weight:
223.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in diethyl ether

density

0.897 g/mL at 25 °C

SMILES string

Cc1cc(C)c([Mg]Br)c(C)c1

InChI

1S/C9H11.BrH.Mg/c1-7-4-8(2)6-9(3)5-7;;/h4-5H,1-3H3;1H;/q;;+1/p-1

InChI key

YXVSITSUDRGILL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Pugh et al.
Nature communications, 6, 7492-7492 (2015-07-02)
Single-molecule magnets are a type of coordination compound that can retain magnetic information at low temperatures. Single-molecule magnets based on lanthanides have accounted for many important advances, including systems with very large energy barriers to reversal of the magnetization, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service