Skip to Content
Merck
All Photos(1)

Documents

639052

Sigma-Aldrich

3,5-Bis(trifluoromethyl)phenylmagnesium bromide solution

0.5 M in THF

Synonym(s):

[3,5-Bis(trifluoromethyl)phenyl]bromomagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4BrF6Mg
CAS Number:
Molecular Weight:
306.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

0.5 M in THF

density

1.015 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cc([Mg]Br)cc(c1)C(F)(F)F

InChI

1S/C8H3F6.BrH.Mg/c9-7(10,11)5-2-1-3-6(4-5)8(12,13)14;;/h2-4H;1H;/q;;+1/p-1

InChI key

FTKNONULORTDRS-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service