Skip to Content
Merck
All Photos(1)

Key Documents

SML2511

Sigma-Aldrich

Levodropropizine

Synonym(s):

Levodropropizine, (-)-Dropropizine, (S)-3-(4-Phenylpiperazin-1-Yl)Propane-1,2-Diol, Levdropropizine, (2S)-3-(4-Phenyl-1-piperazinyl)-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O2
CAS Number:
Molecular Weight:
236.31
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

storage temp.

2-8°C

SMILES string

N1(CCN(CC1)c2ccccc2)C[C@H](O)CO

InChI

1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m0/s1

InChI key

PTVWPYVOOKLBCG-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Levodropropizine has affinity for H1-histaminic and α-adrenergic receptors. Cough suppressant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service