Skip to Content
Merck
All Photos(1)

Key Documents

L0420000

Levodropropizine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2S)-3-(4-Phenyl-1-piperazinyl)-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O2
CAS Number:
Molecular Weight:
236.31
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levodropropizine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1(CCN(CC1)c2ccccc2)C[C@H](O)CO

InChI

1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2/t13-/m0/s1

InChI key

PTVWPYVOOKLBCG-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levodropropizine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung Hee Park et al.
Yonsei medical journal, 54(1), 262-264 (2012-12-12)
We experienced a case of immunoglobulin E (IgE) mediated anaphylaxis to levodropropizine. The patient was an 18-year old Korean woman. After taking the common cold medication including acetaminophen, domperidone, and levodropropizine, skin rash, angioedema and anaphylaxis were developed immediately. As
G Luporini et al.
The European respiratory journal, 12(1), 97-101 (1998-08-13)
Nonproductive cough is a frequent and distressing symptom in patients with lung cancer, and it is not even relieved by palliative chemotherapy. A double-blind, randomized clinical trial regarding the treatment of nonproductive cough was performed in 140 adults with primary
I Yamawaki et al.
European journal of pharmacology, 243(1), 1-6 (1993-10-12)
We investigated the effect of the non-opioid, peripherally acting antitussive agent levodropropizine to reduce neurogenic plasma extravasation in the rat trachea. Levodropropizine (10, 50 and 200 mg/kg) reduced in a dose-dependent manner the extravasation of Evans blue dye evoked by
G Banderali et al.
The Journal of international medical research, 23(3), 175-183 (1995-05-01)
The antitussive efficacy and tolerability of dropropizine and of its enantiomer levodropropizine were evaluated in children with non-productive cough; 258 were evaluable for tolerability and 254 for efficacy. Patients randomly received either 1 mg/kg dropropizine or 2 mg/kg levodropropizine orally
C Bruschi et al.
Pulmonary pharmacology & therapeutics, 16(4), 231-236 (2003-07-10)
To evaluate whether the peripherally acting antitussive levodropropizine could affect the respiratory drive and the breathing pattern, we performed a double-blind, randomised, cross-over trial in 12 healthy volunteers and 12 patients with chronic respiratory impairment associated with chronic obstructive pulmonary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service