Skip to Content
Merck
All Photos(1)

Key Documents

PZ0143

Sigma-Aldrich

PF-4708671

≥98% (HPLC)

Synonym(s):

2-((4-(5-Ethylpyrimidin-4-yl)piperazin-1-yl)methyl)-5-(trifluoromethyl)-1H-benzo[d]imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21F3N6
CAS Number:
Molecular Weight:
390.41
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CCC(C=NC=N1)=C1N2CCN(CC3=NC4=C(C=CC(C(F)(F)F)=C4)N3)CC2

InChI

1S/C19H21F3N6/c1-2-13-10-23-12-24-18(13)28-7-5-27(6-8-28)11-17-25-15-4-3-14(19(20,21)22)9-16(15)26-17/h3-4,9-10,12H,2,5-8,11H2,1H3,(H,25,26)

InChI key

FBLPQCAQRNSVHB-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

PF-4708671 is a selective p70 ribosomal S6 kinase (S6K1) inhibitor. PF-4708671 inhibits S6K1 with a Ki of 20 nM and IC50 of 160 nM, while having no effect on the closely related RSK and MSK kinases.

Features and Benefits

This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wang et al.
International journal of stem cells, 14(3), 341-350 (2021-04-29)
Approximately 15% of couples suffer from infertility worldwide, and male factors contribute to about 30% of total sterility cases. However, there is little progress in treatments due to the obscured understanding of underlying mechanisms. Recently microRNAs have emerged as a
Erika Ishihara et al.
Genes to cells : devoted to molecular & cellular mechanisms, 25(3), 197-214 (2020-01-29)
Cell competition is a biological process by which unfit cells are eliminated from "cell society." We previously showed that cultured mammalian epithelial Madin-Darby canine kidney (MDCK) cells expressing constitutively active YAP were eliminated by apical extrusion when surrounded by "normal"
Mohamed Moustafa-Kamal et al.
Cell reports, 33(1), 108230-108230 (2020-10-08)
mTOR is a serine/threonine kinase and a master regulator of cell growth and proliferation. Raptor, a scaffolding protein that recruits substrates to mTOR complex 1 (mTORC1), is known to be phosphorylated during mitosis, but the significance of this phosphorylation remains

Articles

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service