Skip to Content
Merck
All Photos(1)

Key Documents

Y0000658

Iotrolan for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Iotrolan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48I6N6O18
CAS Number:
Molecular Weight:
1626.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iotrolan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Ic1c(c(c(c(c1C(=O)NC(C(O)CO)CO)I)C(=O)NC(C(O)CO)CO)I)N(C)C(=O)CC(=O)N(C)c2c(c(c(c(c2I)C(=O)NC(C(O)CO)CO)I)C(=O)NC(C(O)CO)CO)I

InChI

1S/C37H48I6N6O18/c1-48(32-28(40)22(34(64)44-12(4-50)16(58)8-54)26(38)23(29(32)41)35(65)45-13(5-51)17(59)9-55)20(62)3-21(63)49(2)33-30(42)24(36(66)46-14(6-52)18(60)10-56)27(39)25(31(33)43)37(67)47-15(7-53)19(61)11-57/h12-19,50-61H,3-11H2,1-2H3,(H,44,64)(H,45,65)(H,46,66)(H,47,67)

InChI key

XUHXFSYUBXNTHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iotrolan for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xenophon Papacharalampous et al.
European journal of radiology, 55(3), 426-430 (2005-09-01)
To examine the effect of intra-articular injection of contrast media, sorbitol and normal saline on the synovial membrane. Sixty three rabbits (126 knees) were used in this study. We injected the knees with amidotrizoate, ioxaglate, iopamidol, iotrol and diluted gadolinium-DTPA
Ingrid Böhm et al.
Investigative radiology, 38(12), 776-783 (2003-11-25)
Mediators of contrast media (CM)-induced allergic/pseudoallergic side effects are largely unknown. We evaluated the possibility of cytokines released as surrogate markers for allergic/pseudoallergic responses in patients who tolerated CM injection well. Interleukin-1beta (IL-1beta), -2, -4, -5, -6; tumor necrosis factor-alpha;
Werner Krause et al.
Investigative radiology, 38(11), 704-711 (2003-10-21)
To develop a new animal model for the evaluation of the renal tolerance of contrast agent and its application to diatrizoate and iotrolan injections in comparison to saline and mannitol solution. The nephrotoxic potential of intravenously injected contrast agents in
Xavier Montet et al.
Surgical and radiologic anatomy : SRA, 27(5), 444-449 (2005-09-01)
The aim of this study was to compare the intra-articular volumes of the wrist's joint measured by CT-arthrography and by dissection on ten cadavers. A good correlation was found between CT-arthrography and dissection in the assessment of the intra-articular volume
Kazuhiro Tsuchiya et al.
European radiology, 13(6), 1438-1443 (2003-05-24)
The advent of a multi-detector row helical CT has made it possible to attain images over a broader area with good spatial resolution. We assessed whether postmyelographic CT scans obtained using this system provided more information than conventional imaging techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service