Skip to Content
Merck
All Photos(1)

Key Documents

Y0000555

Iotrolan

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48I6N6O18
CAS Number:
Molecular Weight:
1626.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iotrolan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C37H48I6N6O18/c1-48(32-28(40)22(34(64)44-12(4-50)16(58)8-54)26(38)23(29(32)41)35(65)45-13(5-51)17(59)9-55)20(62)3-21(63)49(2)33-30(42)24(36(66)46-14(6-52)18(60)10-56)27(39)25(31(33)43)37(67)47-15(7-53)19(61)11-57/h12-19,50-61H,3-11H2,1-2H3,(H,44,64)(H,45,65)(H,46,66)(H,47,67)

InChI key

XUHXFSYUBXNTHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iotrolan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Bender et al.
Neurology, 63(7), 1311-1313 (2004-10-13)
Symptomatic aseptic, chemical meningitis is a rare complication of myelography. Its acute clinical course and standard laboratory findings are indistinguishable from those of bacterial meningitis. The authors present a case of severe postmyelographic chemical meningitis and compare CSF and serum
[Diagnosis and therapy of a spontaneous cerebrospinal fluid leak syndrome].
F Bode et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 181(11), 1087-1089 (2009-10-16)
I Böhm et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 178(3), 306-312 (2006-03-02)
To survey contrast media (CM)-induced alterations of complement factors. In 31 adult patients, who received either an iotrolan (n = 19) or iopromide (n = 12) i. v. injection for CT examination, complement factors C1 q, C3, C4, C5 a
Kwang Su Kim et al.
Anesthesia and analgesia, 108(5), 1658-1661 (2009-04-18)
The purpose of this study was to evaluate epidurography contrast patterns in fluoroscopically guided cervical interlaminar epidural injections using the midline approach. All epidural injections were performed at the C6-7 level in the prone position. Epidurography was performed using a
G Diederichs et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 179(10), 1061-1067 (2007-09-06)
Many patients dislike oral contrast media due to their bad taste. The aim of the present study was to identify a solution that tastes better while providing the same opacification in order to offer oncological patients an alternative to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service